3-Chloro-5,5-dimethyl-2-cyclohexen-1-one 96%

Supplier: Thermo Scientific
Warning

Synonyms: 3-Chloro-5,5-dimethylcyclohex-2-enone

A10728.06 A10728.14 A10728.22
ALFAA10728.06EA 0 SGD
ALFAA10728.06 ALFAA10728.14 ALFAA10728.22
3-Chloro-5,5-dimethyl-2-cyclohexen-1-one 96%
3-Chloro-5,5-dimethyl-2-cyclohexen-1-one

Purity: 96% CAS No.: 17530-69-7 Molecular Formula: C8H11ClO Formula Weight: 158.63 Storage Temperature: 2°C to 8°C Boiling Point: 44-46°/0.1mm Beilstein Reference: 1099253 MDL No.: MFCD00051620

Formula: C₈H₁₁ClO
MW: 158,63 g/mol
Boiling Pt: 44…46 °C (0,1 mmHg)
Density: 1,084
MDL Number: MFCD00051620
CAS Number: 17530-69-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR