DL-Arginine monohydrochloride monohydrate ≥98%

Supplier: Thermo Scientific

A10758.06 A10758.14
ALFAA10758.06EA 0 SGD
ALFAA10758.06 ALFAA10758.14
DL-Arginine monohydrochloride monohydrate ≥98%
DL-Arginine monohydrochloride monohydrate

Purity: 98+% CAS No.: 332360-01-7 Molecular Formula: C6H14N4O2.HCl.H2O Formula Weight: 228.69 (210.67anhy) Beilstein Reference: 4158960 MDL No.: MFCD00151031

Formula: C₆H₁₄N₄O₂·HCl·H₂O
Storage Temperature: Ambient
MDL Number: MFCD00151031
CAS Number: 332360-01-7
EINECS: 250-903-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR