1-(Phenylsulphonyl)pyrrole 98%
Supplier: Thermo Scientific
|
Warning
|
A11599.03
A11599.06
A11599.14
ALFAA11599.03EA
0
SGD
ALFAA11599.03
ALFAA11599.06
ALFAA11599.14
1-(Phenylsulphonyl)pyrrole 98%
1-(Phenylsulphonyl)pyrrole
Purity: 98%
CAS No.: 16851-82-4
Molecular Formula: C10H9NO2S
Formula Weight: 207.25
Melting Point: 87-90°
Beilstein Reference: 1619427
MDL No.: MFCD00067739
Formula:
C₁₀H₉NO₂S MW: 207,247 g/mol Melting Pt: 87…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00067739 CAS Number: 16851-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...