1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one 96%
Supplier: Thermo Scientific
Synonyms:
2,4-Dihydro-2-methyl-5-(trifluoromethyl)-3H-pyrazol-3-one
A11890.06
A11890.14
ALFAA11890.06EA
0
SGD
ALFAA11890.06
ALFAA11890.14
1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one 96%
1-Methyl-3-trifluoromethyl-2-pyrazolin-5-one
Purity: 96%
CAS No.: 1481-02-3
Molecular Formula: C5H5F3N2O
Formula Weight: 166.10
Melting Point: 178-180°
Beilstein Reference: 5811070
MDL No.: MFCD00051654
Formula:
C₅H₅F₃N₂O MW: 166,1 g/mol Melting Pt: 178…180 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051654 CAS Number: 1481-02-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...