Tetraphenylphthalic anhydride ≥97%

Supplier: Thermo Scientific
Warning

A11998.06 A11998.14 A11998.09
ALFAA11998.06EA 0 SGD
ALFAA11998.06 ALFAA11998.14 ALFAA11998.09
Tetraphenylphthalic anhydride ≥97%
Tetraphenylphthalic anhydride

Purity: 98% CAS No.: 4741-53-1 Molecular Formula: C32H20O3 Formula Weight: 452.51 Melting Point: 289-297° Beilstein Reference: 353504 MDL No.: MFCD00044504

Formula: C₃₂H₂₀O₃
MW: 452,51 g/mol
Melting Pt: 289…297 °C
Storage Temperature: Ambient
MDL Number: MFCD00044504
CAS Number: 4741-53-1
EINECS: 225-253-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR