4-Hydroxy-3-nitrocoumarin 98%
Supplier: Thermo Scientific
Synonyms:
4-Hydroxy-3-nitro-2H-chromen-2-one
A12221.06
A12221.14
ALFAA12221.06EA
0
SGD
ALFAA12221.06
ALFAA12221.14
4-Hydroxy-3-nitrocoumarin 98%
4-Hydroxy-3-nitrocoumarin
Purity: 98%
CAS No.: 20261-31-8
Molecular Formula: C9H5NO5
Formula Weight: 207.14
Melting Point: ca 171° dec.
Beilstein Reference: 1247343
MDL No.: MFCD00051671
Formula:
C₉H₅NO₅ MW: 207,14 g/mol Melting Pt: ∼ 171 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051671 CAS Number: 20261-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...