1,4-Dioxa-8-azaspiro[4.5]decane ≥98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
1,4-Dioxa-8-azaspiro[4.5]decane, 4-Piperidone ethyleneketal
A12419.09
A12419.18
A12419.30
ALFAA12419.09EA
0
SGD
ALFAA12419.09
ALFAA12419.18
ALFAA12419.30
1,4-Dioxa-8-azaspiro[4.5]decane ≥98%
1,4-Dioxa-8-azaspiro[4.5]decane
Purity: 98+%
CAS No.: 177-11-7
Molecular Formula: C7H13NO2
Formula Weight: 143.19
Flash Point: 81°(177°F)
Refractive Index: 1.4819
Boiling Point: 99-101°/14mm
Beilstein Reference: 506799
MDL No.: MFCD00005976
Formula:
C₇H₁₃NO₂ MW: 143,186 g/mol Boiling Pt: 99…101 °C (14 mmHg) Density: 1,117 Flash Pt: 81 °C (177 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005976 CAS Number: 177-11-7 EINECS: 205-868-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...