Methyl-2-(4-(tert-butyl)phenyl)acetate 96%
Supplier: Thermo Scientific
Synonyms:
Methyl 2-(4-(tert-butyl)phenyl)acetate
A12421.06
A12421.14
ALFAA12421.06EA
0
SGD
ALFAA12421.06
ALFAA12421.14
Methyl-2-(4-(tert-butyl)phenyl)acetate 96%
Methyl-2-(4-(tert-butyl)phenyl)acetate
Purity: 97%
CAS No.: 3549-23-3
Molecular Formula: C13H18O2
Formula Weight: 206.29
Flash Point:
Formula:
C₁₃H₁₈O₂ MW: 206,29 g/mol Boiling Pt: 149…151 °C (30 mmHg) Density: 0,994 Flash Pt: >100 °C (212 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00051913 CAS Number: 3549-23-3 EINECS: 222-602-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...