Tri(2-thienyl)phosphine ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Tri(thiophen-2-yl)phosphine

A13379.03 A13379.06 A13379.14
ALFAA13379.03EA 0 SGD
ALFAA13379.03 ALFAA13379.06 ALFAA13379.14
Tri(2-thienyl)phosphine ≥98%
Tri(2-thienyl)phosphine

Purity: 98+% CAS No.: 24171-89-9 Molecular Formula: C12H9PS3 Formula Weight: 280.37 Odour: Recognizable Melting Point: 28-30° Beilstein Reference: 203736 MDL No.: MFCD00015471

Formula: C₁₂H₉PS₃
MW: 280,36 g/mol
Melting Pt: 28…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00015471
CAS Number: 24171-89-9
EINECS: 246-059-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR