Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate 98%
Supplier: Thermo Scientific
Synonyms:
Ethyl 4-methyl-1,2,3-thiadiazole-5-carboxylate
A13399.03
A13399.06
ALFAA13399.03EA
0
SGD
ALFAA13399.03
ALFAA13399.06
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate 98%
Ethyl-4-methyl-1,2,3-thiadiazole-5-carboxylate
Purity: 98%
CAS No.: 18212-20-9
Molecular Formula: C6H8N2O2S
Formula Weight: 172.21
Refractive Index: 1.5040
Boiling Point: 117-118°/15mm
Beilstein Reference: 609140
MDL No.: MFCD00052549
Formula:
C₆H₈N₂O₂S MW: 172,21 g/mol Boiling Pt: 117…118 °C (15 mmHg) Density: 1,265 Storage Temperature: Ambient |
MDL Number:
MFCD00052549 CAS Number: 18212-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...