Naphthalene-2-sulphonic acid hydrate 94%

Supplier: Thermo Scientific
Danger

Synonyms: 2-Naphthalenesulphonic acid hydrate

A13445.0B A13445.18 A13445.30
ALFAA13445.0BEA 0 SGD
ALFAA13445.0B ALFAA13445.18 ALFAA13445.30
Naphthalene-2-sulphonic acid hydrate 94%
Naphthalene-2-sulphonic acid hydrate

Purity: 94% CAS No.: 76530-12-6 Molecular Formula: C10H8O3S.xH2O Formula Weight: 208.23(anhy) UN No.: UN2585 MDL No.: MFCD00149271

Formula: C₁₀H₈O₃S·xH₂O
MW: 208,231 g/mol (anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149271
CAS Number: 76530-12-6
ADR: 8,III
Merck Index: 14,6377

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR