N-Carbamoylmaleimide 90%, tech.

Supplier: Thermo Scientific
Warning

A13685.09 A13685.18
ALFAA13685.09EA 0 SGD
ALFAA13685.09 ALFAA13685.18
N-Carbamoylmaleimide 90%, tech.
N-Carbamoylmaleimide

Purity: 90% CAS No.: 3345-50-4 Molecular Formula: C5H4N2O3 Formula Weight: 140.10 Melting Point: 157-160° Beilstein Reference: 117919 MDL No.: MFCD00014539

Formula: C₅H₄N₂O₃
MW: 140,10 g/mol
Melting Pt: 157…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00014539
CAS Number: 3345-50-4
EINECS: 222-097-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR