6-Chloro-5-fluoro-2-mercaptobenzimidazole 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
6-Chloro-5-fluorobenzimidazole-2-thiol
A15367.03
A15367.06
ALFAA15367.03EA
0
SGD
ALFAA15367.03
ALFAA15367.06
6-Chloro-5-fluoro-2-mercaptobenzimidazole 98%
6-Chloro-5-fluoro-2-mercaptobenzimidazole
Purity: 98%
CAS No.: 142313-30-2
Molecular Formula: C7H4ClFN2S
Formula Weight: 202.64
Melting Point: ca 308° dec.
MDL No.: MFCD00052490
Formula:
C₇H₄ClFN₂S MW: 202,64 g/mol Melting Pt: ∼ 308 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00052490 CAS Number: 142313-30-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...