2-(Pyridin-3-yl)acetic acid hydrochloride ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: Pyridyl-3-acetic acid hydrochloride, 3-Pyridylacetic acid hydrochloride

A15599.06 A15599.14 A15599.22
ALFAA15599.06EA 0 SGD
ALFAA15599.06 ALFAA15599.14 ALFAA15599.22
2-(Pyridin-3-yl)acetic acid hydrochloride ≥98%
2-(Pyridin-3-yl)acetic acid hydrochloride

Purity: 98+% CAS No.: 6419-36-9 Molecular Formula: C7H7NO2.HCl Formula Weight: 173.60 Melting Point: 158-162° Beilstein Reference: 3696630 MDL No.: MFCD00012819

Formula: C₇H₈ClNO₂
MW: 173,60 g/mol
Melting Pt: 158…162 °C
Storage Temperature: Ambient
MDL Number: MFCD00012819
CAS Number: 6419-36-9
EINECS: 229-148-6
Merck Index: 14,7971

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR