trans,trans-2,4-Decadien-1-ol 90% remainder mainly trans, cis isomer
Supplier: Thermo Scientific
A15989.03
A15989.06
A15989.14
ALFAA15989.03EA
0
SGD
ALFAA15989.03
ALFAA15989.06
ALFAA15989.14
trans,trans-2,4-Decadien-1-ol 90% remainder mainly trans, cis isomer
trans,trans-2,4-Decadien-1-ol
Purity: 90%
CAS No.: 18409-21-7
Molecular Formula: C10H18O
Formula Weight: 154.26
Flash Point:
Formula:
C₁₀H₁₈O MW: 154,26 g/mol Boiling Pt: 80 °C (0,5 mmHg) Density: 0,870 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014052 CAS Number: 18409-21-7 EINECS: 242-291-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...