Diphenyl-N-cyanocarbonimidate 97%

Supplier: Thermo Scientific
Danger

Synonyms: Diphenyl N-cyanocarbonimidate, N-Cyanocarbonimidic Acid Diphenyl Ester

A16600.06 A16600.14
ALFAA16600.06EA 0 SGD
ALFAA16600.06 ALFAA16600.14
Diphenyl-N-cyanocarbonimidate 97%
Diphenyl-N-cyanocarbonimidate

Purity: 97% CAS No.: 79463-77-7 Molecular Formula: C14H10N2O2 Formula Weight: 238.25 Melting Point: 155-158° MDL No.: MFCD00010380

Formula: C₁₄H₁₀N₂O₂
MW: 238,25 g/mol
Melting Pt: 155…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00010380
CAS Number: 79463-77-7
EINECS: 427-300-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR