8-Ethoxyquinoline-5-sulphonic acid sodium salt hydrate 97%

Supplier: Thermo Scientific

A16668.18 A16668.30
ALFAA16668.18EA 0 SGD
ALFAA16668.18 ALFAA16668.30
8-Ethoxyquinoline-5-sulphonic acid sodium salt hydrate 97%
8-Ethoxyquinoline-5-sulphonic acid sodium salt hydrate

Purity: 97% CAS No.: 80789-76-0 Molecular Formula: C11H10NNaO4S.xH2O Formula Weight: 275.26(anhy) Melting Point: 280° MDL No.: MFCD00149478

Formula: C₁₁H₁₀NNaO₄S·xH₂O
MW: 275,26 g/mol (anhy)
Melting Pt: >280 °C
Storage Temperature: Ambient
MDL Number: MFCD00149478
CAS Number: 80789-76-0
EINECS: 230-651-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR