3-(2-Naphthyl)acrylic acid 98%

Supplier: Thermo Scientific
Warning

Synonyms: 3-(Naphthalen-2-yl)acrylic acid

A16715.03 A16715.06
ALFAA16715.03EA 83.8 SGD
ALFAA16715.03 ALFAA16715.06
3-(2-Naphthyl)acrylic acid 98%
3-(2-Naphthyl)acrylic acid

Purity: 98% CAS No.: 51557-26-7 Molecular Formula: C13H10O2 Formula Weight: 198.22 MDL No.: MFCD00209666

Formula: C₁₃H₁₀O₂
MW: 198,22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00209666
CAS Number: 51557-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR