Lobeline sulphate 99%

Supplier: Thermo Fisher Scientific
Danger

A17326.03 A17326.06
ALFAA17326.03EA 0 SGD
ALFAA17326.03 ALFAA17326.06
Lobeline sulphate 99%
Lobeline sulphate

Purity: 99% CAS No.: 134-64-5 Molecular Formula: C44H54N2O4.H2SO4 Formula Weight: 773.01 UN No.: UN1544 MDL No.: MFCD00135592

Formula: C₄₄H₅₄N₂O₄·H₂SO₄
MW: 773.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00135592
CAS Number: 134-64-5
EINECS: 205-151-8
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR