Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy 99% free radical
Supplier: Thermo Scientific
A17381.03
A17381.06
ALFAA17381.03EA
0
SGD
ALFAA17381.03
ALFAA17381.06
Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy 99% free radical
Carbamoyl-2,2,5,5-tetramethyl-3-pyrrolin-1-yloxy
Purity: 99%
CAS No.: 3229-73-0
Molecular Formula: C9H15N2O2
Formula Weight: 183.23
Storage Temperature: 2°C to 8°C
Melting Point: 198-200°
Beilstein Reference: 4138020
MDL No.: MFCD00005341
Formula:
C₉H₁₅N₂O₂ MW: 183,23 g/mol Melting Pt: 198…200 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005341 CAS Number: 3229-73-0 EINECS: 221-765-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...