Hexafluoro-2-(p-tolyl)isopropanol 97%

Supplier: Thermo Scientific

Synonyms: 1,1,1,3,3,3-Hexafluoro-2-(4-methylphenyl)propan-2-ol

A18123.06 A18123.14
ALFAA18123.06EA 0 SGD
ALFAA18123.06 ALFAA18123.14
Hexafluoro-2-(p-tolyl)isopropanol 97%
Hexafluoro-2-(p-tolyl)isopropanol

Purity: 97% CAS No.: 2010-61-9 Molecular Formula: C10H8F6O Formula Weight: 258.17 Refractive Index: 1.4240 Boiling Point: 181-182° Beilstein Reference: 2127760 MDL No.: MFCD00015357

Formula: C₁₀H₈F₆O
MW: 258,17 g/mol
Boiling Pt: 181…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00015357
CAS Number: 2010-61-9
EINECS: 217-928-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR