Scandium(III) acetate hydrate 99.9% (metals basis)

Supplier: Thermo Scientific

A18271.06 A18271.03
ALFAA18271.06EA 0 SGD
ALFAA18271.06 ALFAA18271.03
Scandium(III) acetate hydrate 99.9% (metals basis)
Scandium(III) acetate hydrate

Purity: 99.9% CAS No.: 304675-64-7 Molecular Formula: C6H9O6Sc.xH2O Formula Weight: 222.10(anhy) MDL No.: MFCD00013043

Formula: C₆H₉O₆Sc·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00013043
CAS Number: 304675-64-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR