Ethyl-3-methyl-4H-1,4-benzothiazine-2-carboxylate 98%

Supplier: Thermo Scientific
Warning

A18360.03 A18360.06
ALFAA18360.03EA 0 SGD
ALFAA18360.03 ALFAA18360.06
Ethyl-3-methyl-4H-1,4-benzothiazine-2-carboxylate 98%
Ethyl-3-methyl-4H-1,4-benzothiazine-2-carboxylate

Purity: 98% CAS No.: 7625-01-6 Molecular Formula: C12H13NO2S Formula Weight: 235.31 Melting Point: 142-144° MDL No.: MFCD00173950

Formula: C₁₂H₁₃NO₂S
MW: 235,31 g/mol
Melting Pt: 142…144 °C
Storage Temperature: Ambient
MDL Number: MFCD00173950
CAS Number: 7625-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR