O(1),O(3)-Bis(carboxymethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene

Supplier: Thermo Scientific

A19192.77 A19192.02
ALFAA19192.77EA 0 SGD
ALFAA19192.77 ALFAA19192.02
O(1),O(3)-Bis(carboxymethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene
O(1),O(3)-Bis(carboxymethyl)-O(2),O(4)-dimethyl-p-tert-butylcalix[4]arene

CAS No.: 136157-98-7 Molecular Formula: C50H64O8 Formula Weight: 793.06 Melting Point: 250-252° MDL No.: MFCD00798565

Formula: C₅₀H₆₄O₈
MW: 793,06 g/mol
Melting Pt: 250…252 °C
Storage Temperature: Ambient
MDL Number: MFCD00798565
CAS Number: 136157-98-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR