Glycidyl-4-methoxyphenyl ether 98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-((4-Methoxyphenoxy)methyl)oxirane

A19780.14 A19780.22 A19780.06
ALFAA19780.14EA 0 SGD
ALFAA19780.14 ALFAA19780.22 ALFAA19780.06
Glycidyl-4-methoxyphenyl ether 98%
Glycidyl-4-methoxyphenyl ether

Purity: 98% CAS No.: 2211-94-1 Molecular Formula: C10H12O3 Formula Weight: 180.20 Flash Point: 110°(230°F) Melting Point: 45-48° MDL No.: MFCD00005135

Formula: C₁₀H₁₂O₃
MW: 180,20 g/mol
Melting Pt: 45…48 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005135
CAS Number: 2211-94-1
EINECS: 218-653-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR