(+)-3-Bromocamphor-8-sulphonic acid ammonium salt 98%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
Ammonium ((1R,3S,4S,7R)-3-bromo-1,7-dimethyl-2-oxobicyclo[2.2.1]heptan-7-yl)methanesulphonate
A19886.06
A19886.14
A19886.22
ALFAA19886.06EA
0
SGD
ALFAA19886.06
ALFAA19886.14
ALFAA19886.22
(+)-3-Bromocamphor-8-sulphonic acid ammonium salt 98%
(+)-3-Bromocamphor-8-sulphonic acid ammonium salt
Purity: 98%
CAS No.: 14575-84-9
Molecular Formula: C10H18BrNO4S
Formula Weight: 328.23
Melting Point: ca 284° dec.
MDL No.: MFCD00167438
Formula:
C₁₀H₁₈BrNO₄S MW: 328,22 g/mol Melting Pt: ∼ 284 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00167438 CAS Number: 14575-84-9 EINECS: 238-616-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...