2-Bromo-1-chloro-2-methylpropane ≥90%

Supplier: Thermo Scientific
Warning

A19974.06 A19974.14
ALFAA19974.06EA 91.4 SGD
ALFAA19974.06 ALFAA19974.14
2-Bromo-1-chloro-2-methylpropane ≥90%
2-Bromo-1-chloro-2-methylpropane

Purity: 90+% CAS No.: 2074-80-8 Molecular Formula: Br(CH3)2CCH2Cl Formula Weight: 171.47 Flash Point: 37°(98°F) Refractive Index: 1.4740 Odour: Aromatic Boiling Point: 127-128° UN No.: UN1993 MDL No.: MFCD00049143

Formula: Br(CH₃)₂CCH₂Cl
MW: 171,47 g/mol
Boiling Pt: 127…128 °C
Density: 1,44
Flash Pt: 37 °C (98 °F)
Storage Temperature: Ambient
MDL Number: MFCD00049143
CAS Number: 2074-80-8
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR