Methyl-3-(thien-2-yl)acrylate 97%

Supplier: Thermo Scientific
Warning

Synonyms: Methyl 3-(thiophen-2-yl)acrylate

B20177.03 B20177.06
ALFAB20177.03EA 0 SGD
ALFAB20177.03 ALFAB20177.06
Methyl-3-(thien-2-yl)acrylate 97%
Methyl-3-(thien-2-yl)acrylate

Purity: 97% CAS No.: 20883-96-9 Molecular Formula: C8H8O2S Formula Weight: 168.22 Melting Point: 48-49° MDL No.: MFCD00173772

Formula: C₈H₈O₂S
MW: 168,22 g/mol
Melting Pt: 48…49 °C
Storage Temperature: Ambient
MDL Number: MFCD00173772
CAS Number: 20883-96-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR