Dimethallyl carbonate 97%
Supplier: Thermo Scientific
Synonyms:
Carbonic acid dimethallyl ester, Bis(2-methylallyl) carbonater
B20414.06
B20414.14
ALFAB20414.06EA
0
SGD
ALFAB20414.06
ALFAB20414.14
Dimethallyl carbonate 97%
Dimethallyl carbonate
Purity: 97%
CAS No.: 64057-79-0
Molecular Formula: C11H18O3
Formula Weight: 170.21
Boiling Point: 199-202°
MDL No.: MFCD00008598
Formula:
C₉H₁₄O₃ MW: 170,208 g/mol Boiling Pt: 199…202 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008598 CAS Number: 64057-79-0 EINECS: 264-645-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...