5-Methyl-1,3-benzoxazole-2-thiol 97%
Supplier: Thermo Scientific
|
Warning
|
Synonyms:
5-Methyl-2,3-dihydro-1,3-benzoxazole-2-thione
B20515.03
B20515.06
ALFAB20515.03EA
0
SGD
ALFAB20515.03
ALFAB20515.06
5-Methyl-1,3-benzoxazole-2-thiol 97%
5-Methyl-1,3-benzoxazole-2-thiol
Purity: 97%
CAS No.: 22876-22-8
Molecular Formula: C8H7NOS
Formula Weight: 165.21
Melting Point: 223-225°
MDL No.: MFCD00052492
Formula:
C₈H₇NOS MW: 165,22 g/mol Melting Pt: 223…225 °C Storage Temperature: Ambient |
MDL Number:
MFCD08447019 CAS Number: 22876-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...