3-Chloroadamantane-1-carboxylic acid 97%

Supplier: Thermo Scientific
Warning

B20543.03 B20543.06
ALFAB20543.03EA 0 SGD
ALFAB20543.03 ALFAB20543.06
3-Chloroadamantane-1-carboxylic acid 97%
3-Chloroadamantane-1-carboxylic acid

Purity: 97% CAS No.: 34859-74-0 Molecular Formula: C11H15ClO2 Formula Weight: 214.69 MDL No.: MFCD00167831

Formula: C₁₁H₁₅ClO₂
Storage Temperature: Ambient
MDL Number: MFCD00167831
CAS Number: 34859-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR