4-Acetyl-4'-nitrodiphenyl ether 97%
Supplier: Thermo Scientific
B20592.03
B20592.06
ALFAB20592.03EA
0
SGD
ALFAB20592.03
ALFAB20592.06
4-Acetyl-4'-nitrodiphenyl ether 97%
4-Acetyl-4'-nitrodiphenyl ether
Purity: 97%
CAS No.: 75919-92-5
Molecular Formula: C14H11NO4
Formula Weight: 257.25
Melting Point: 79-81°
Beilstein Reference: 2135124
MDL No.: MFCD00052956
Formula:
C₁₄H₁₁NO₄ MW: 257,25 g/mol Melting Pt: 79…81 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052956 CAS Number: 75919-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...