4-Methyl-1H-imidazole-5-carbaldehyde 98%

Supplier: Thermo Scientific
Warning

Synonyms: 4-Methyl-5-imidazolecarboxaldehyde

B20608.03 B20608.06
ALFAB20608.03EA 0 SGD
ALFAB20608.03 ALFAB20608.06
4-Methyl-1H-imidazole-5-carbaldehyde 98%
4-Methyl-1H-imidazole-5-carbaldehyde

Purity: 99% CAS No.: 68282-53-1 Molecular Formula: C5H6N2O Formula Weight: 110.12 Melting Point: 166-170° MDL No.: MFCD00173728

Formula: C₅H₆N₂O
MW: 110,12 g/mol
Melting Pt: 166…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00173728
CAS Number: 68282-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR