2-Phenyl-2-pentanol 98%
Supplier: Thermo Scientific
Synonyms:
2-Phenylpentan-2-ol
B20854.14
B20854.03
ALFAB20854.14EA
0
SGD
ALFAB20854.14
ALFAB20854.03
2-Phenyl-2-pentanol 98%
2-Phenyl-2-pentanol
Purity: 98%
CAS No.: 4383-18-0
Molecular Formula: C11H16O
Formula Weight: 164.25
MDL No.: MFCD00046592
Formula:
C₁₁H₁₆O MW: 164,25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00046592 CAS Number: 4383-18-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...