2-Phenyl-1H-indole-3-carbaldehyde 97%

Supplier: Thermo Scientific

Synonyms: 3-Formyl-2-phenylindole, 2-Phenylindole-3-carboxaldehyde

B21041.03 B21041.06
ALFAB21041.03EA 75 SGD
ALFAB21041.03 ALFAB21041.06
2-Phenyl-1H-indole-3-carbaldehyde 97%
2-Phenyl-1H-indole-3-carbaldehyde

Purity: 97% CAS No.: 25365-71-3 Molecular Formula: C15H11NO Formula Weight: 221.26 Melting Point: 249-253° MDL No.: MFCD00435481

Formula: C₁₅H₁₁NO
MW: 221,26 g/mol
Melting Pt: 249…253 °C
Storage Temperature: Ambient
MDL Number: MFCD00435481
CAS Number: 25365-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR