2-Phenylethyl propionate 97%

Supplier: Thermo Scientific

Synonyms: Propionic Acid Phenethyl Ester, Propionic Acid 2-Phenylethyl Ester, 2-Phenylethyl Propionate, Phenethyl Propionate

B21283.18 B21283.30
ALFAB21283.18EA 0 SGD
ALFAB21283.18 ALFAB21283.30
2-Phenylethyl propionate 97%
2-Phenylethyl propionate

Purity: 97% CAS No.: 122-70-3 Molecular Formula: C6H5(CH2)2OCOC2H5 Formula Weight: 178.22 Flash Point: 106°(223°F) Refractive Index: 1.4940 Odour: Floral Boiling Point: 245° MDL No.: MFCD00027008

Formula: C₁₁H₁₄O₂
MW: 178,23 g/mol
Boiling Pt: 245 °C
Density: 1,007
Flash Pt: 106 °C (223 °F)
Storage Temperature: Ambient
MDL Number: MFCD00027008
CAS Number: 122-70-3
EINECS: 204-567-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR