Bis(2-bromobenzyl)amine hydrochloride 99%

Supplier: Thermo Scientific
Warning

B21318.06 B21318.14 B21318.22
ALFAB21318.06EA 0 SGD
ALFAB21318.06 ALFAB21318.14 ALFAB21318.22
Bis(2-bromobenzyl)amine hydrochloride 99%
Bis(2-bromobenzyl)amine hydrochloride

Purity: 99% CAS No.: 336615-45-3 Molecular Formula: C14H13Br2N.HCl Formula Weight: 391.53 Melting Point: 165-168° MDL No.: MFCD01632133

Formula: C₁₄H₁₃Br₂N·HCl
MW: 391,53 g/mol
Melting Pt: 165…168 °C
Storage Temperature: Ambient
MDL Number: MFCD01632133
CAS Number: 336615-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR