Piperonyl isobutyrate 99%

Supplier: Thermo Scientific

B21321.14 B21321.22
ALFAB21321.14EA 0 SGD
ALFAB21321.14 ALFAB21321.22
Piperonyl isobutyrate 99%
Piperonyl isobutyrate

Purity: 99% CAS No.: 5461-08-5 Molecular Formula: C12H14O4 Formula Weight: 222.24 Flash Point: 110°(230°F) Refractive Index: 1.5120 Boiling Point: 91-92°/0.005mm MDL No.: MFCD00005826

Formula: C₁₂H₁₄O₄
Boiling Pt: 91…92 °C (0,005 mmHg)
Density: 1,154
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005826
CAS Number: 5461-08-5
EINECS: 226-745-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR