1-Acetylindole-3-carboxaldehyde 98%

Supplier: Thermo Scientific
Warning

Synonyms: 1-Acetyl-3-indolecarboxaldehyde, 1-Acetyl-1H-indole-3-carbaldehyde

B21527.03 B21527.06 B21527.14
ALFAB21527.03EA 0 SGD
ALFAB21527.03 ALFAB21527.06 ALFAB21527.14
1-Acetylindole-3-carboxaldehyde 98%
1-Acetylindole-3-carboxaldehyde

Purity: 98% CAS No.: 22948-94-3 Molecular Formula: C11H9NO2 Formula Weight: 187.20 Melting Point: 160-164° MDL No.: MFCD00039691

Formula: C₁₁H₉NO₂
MW: 187,20 g/mol
Melting Pt: 160…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00039691
CAS Number: 22948-94-3
EINECS: 245-347-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR