n-Octyl-L-2-pyrrolidinone-5-carboxylate 98%
Supplier: Thermo Scientific
B21838.06
B21838.14
ALFAB21838.06EA
0
SGD
ALFAB21838.06
ALFAB21838.14
n-Octyl-L-2-pyrrolidinone-5-carboxylate 98%
n-Octyl-L-2-pyrrolidinone-5-carboxylate
Purity: 98%
CAS No.: 4931-70-8
Molecular Formula: C13H23NO3
Formula Weight: 241.33
Refractive Index: 1.4720
Boiling Point: 140°/0.06mm
Beilstein Reference: 478315
MDL No.: MFCD00236053
Formula:
C₁₃H₂₃NO₃ MW: 241,33 g/mol Boiling Pt: 140 °C (0,06 mmHg) Storage Temperature: Ambient |
MDL Number:
MFCD00236053 CAS Number: 4931-70-8 EINECS: 225-569-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...