5,5-Dimethyl-1,3-dioxane-2-butanal 96%

Supplier: Thermo Scientific
Warning

B21858.09 B21858.88
ALFAB21858.09EA 0 SGD
ALFAB21858.09 ALFAB21858.88
5,5-Dimethyl-1,3-dioxane-2-butanal 96%
5,5-Dimethyl-1,3-dioxane-2-butanal

Purity: 96% CAS No.: 127600-13-9 Molecular Formula: C10H18O3 Formula Weight: 186.25 Refractive Index: 1.4490 MDL No.: MFCD02683076

Formula: C₁₀H₁₈O₃
MW: 186,25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02683076
CAS Number: 127600-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR