1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
Supplier: Thermo Scientific
Synonyms:
1-Benzyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 1-Benzyl Ester, BOC-L-Aspartic acid 1-benzyl ester, N-Boc-L-aspartic Acid 1-Benzyl Ester, Boc-Asp-OBzl, Boc-Asp-Obzl
B22314.03
B22314.06
ALFAB22314.03EA
66.8
SGD
ALFAB22314.03
ALFAB22314.06
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate
Purity: 99%
CAS No.: 30925-18-9
Molecular Formula: C16H21NO6
Formula Weight: 323.35
Melting Point: 95-97°
MDL No.: MFCD00065563
Formula:
C₁₆H₂₁NO₆ MW: 323,35 g/mol Melting Pt: 95…97 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065563 CAS Number: 30925-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...