2-Carboxyphenyl salicylate 98%

Supplier: Thermo Scientific
Warning

Synonyms: 2-Salicyloyloxybenzoic Acid, O-Salicyloylsalicylic Acid, Salicylic Acid 2-Carboxyphenyl Ester, Salsalate

B22444.14 B22444.22 B22444.36
ALFAB22444.14EA 0 SGD
ALFAB22444.14 ALFAB22444.22 ALFAB22444.36
2-Carboxyphenyl salicylate 98%
2-Carboxyphenyl salicylate

Purity: 98% CAS No.: 552-94-3 Molecular Formula: C14H10O5 Formula Weight: 258.23 Melting Point: 132-138° MDL No.: MFCD00020252

Formula: C₁₄H₁₀O₅
MW: 258,23 g/mol
Melting Pt: 132…138 °C
Storage Temperature: Ambient
MDL Number: MFCD00020252
CAS Number: 552-94-3
EINECS: 209-027-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR