4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone ≥98%

Supplier: Thermo Scientific

Synonyms: 4-Benzoyl-5-methyl-2-phenyl-2,4-dihydro-3H-pyrazol-3-one

B22530.09 B22530.18
ALFAB22530.09EA 0 SGD
ALFAB22530.09 ALFAB22530.18
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone ≥98%
4-Benzoyl-3-methyl-1-phenyl-5-pyrazolone

Purity: 98+% CAS No.: 4551-69-3 Molecular Formula: C17H14N2O2 Formula Weight: 278.31 Melting Point: 90-92° Beilstein Reference: 888605 MDL No.: MFCD00003136

Formula: C₁₇H₁₄N₂O₂
MW: 278,31 g/mol
Melting Pt: 90…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00003136
CAS Number: 4551-69-3
EINECS: 224-918-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR