2,4-Dimethyl-2-pentenoic acid, predominantly trans 99%
Supplier: Thermo Scientific
|
Danger
|
B22701.09
B22701.88
ALFAB22701.09EA
0
SGD
ALFAB22701.09
ALFAB22701.88
2,4-Dimethyl-2-pentenoic acid, predominantly trans 99%
2,4-Dimethyl-2-pentenoic acid, predominantly trans
Purity: 99%
CAS No.: 66634-97-7
Molecular Formula: C7H12O2
Formula Weight: 128.17
Refractive Index: 1.4540
UN No.: UN3265
MDL No.: MFCD00209505
Formula:
C₇H₁₂O₂ MW: 128,17 g/mol Density: 0,949 Storage Temperature: Ambient |
MDL Number:
MFCD00209505 CAS Number: 66634-97-7 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...