cis-Dichlorobis(pyridine)platinum(II) ≥99%

Supplier: Thermo Scientific
Warning

B22865.01 B22865.03
ALFAB22865.01EA 0 SGD
ALFAB22865.01 ALFAB22865.03
cis-Dichlorobis(pyridine)platinum(II) ≥99%
cis-Dichlorobis(pyridine)platinum(II)

Purity: 99% CAS No.: 14872-21-0 Molecular Formula: C10H10Cl2N2Pt Formula Weight: 424.19 Melting Point: 282-288° MDL No.: MFCD00058858

Formula: C₁₀H₁₀Cl₂N₂Pt
MW: 424,19 g/mol
Melting Pt: 282…288 °C
Storage Temperature: Ambient
MDL Number: MFCD00058858
CAS Number: 15227-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR