Ethyl-1-adamantanecarboxylate ≥98%

Supplier: Thermo Fisher Scientific

B24518.04 B24518.09
ALFAB24518.04EA 0 SGD
ALFAB24518.04 ALFAB24518.09
Ethyl-1-adamantanecarboxylate ≥98%
Ethyl-1-adamantanecarboxylate

Purity: 98+% CAS No.: 2094-73-7 Molecular Formula: C13H20O2 Formula Weight: 208.30 Refractive Index: 1.4890 Boiling Point: 90-91°/1mm Beilstein Reference: 2050027 MDL No.: MFCD00078272

Formula: C₁₃H₂₀O₂
Boiling Pt: 90…91 °C (1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00078272
CAS Number: 2094-73-7
EINECS: 218-253-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR