3-Methylbenzo[f]quinoline 97%

Supplier: Thermo Scientific
Warning

B24920.06 B24920.14 B24920.22
ALFAB24920.06EA 0 SGD
ALFAB24920.06 ALFAB24920.14 ALFAB24920.22
3-Methylbenzo[f]quinoline 97%
3-Methylbenzo[f]quinoline

Purity: 97% CAS No.: 85-06-3 Molecular Formula: C14H11N Formula Weight: 193.25 Melting Point: 73-78° MDL No.: MFCD00046906

Formula: C₁₄H₁₁N
MW: 193,25 g/mol
Melting Pt: 73…78 °C
Storage Temperature: Ambient
MDL Number: MFCD00046906
CAS Number: 85-06-3
EINECS: 201-584-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR