N-Carbobenzyloxy-D-proline ≥98%
Supplier: Thermo Scientific
Synonyms:
(R)-1-((Benzyloxy)carbonyl)pyrrolidine-2-carboxylic acid, Z-D-Pro-OH, N-Cbz-D-proline, Cbz-D-Pro-OH, ((Benzyloxy)carbonyl)-D-proline, N-Benzyloxycarbonyl-D-proline
B25005.06
B25005.14
ALFAB25005.06EA
0
SGD
ALFAB25005.06
ALFAB25005.14
N-Carbobenzyloxy-D-proline ≥98%
N-Carbobenzyloxy-D-proline
Purity: 98+%
CAS No.: 6404-31-5
Molecular Formula: C13H15NO4
Formula Weight: 249.27
Storage Temperature: 2°C to 8°C
Melting Point: 76-78°
Beilstein Reference: 485188
Optical Rotation: +40° (c=2 in ethanol)
MDL No.: MFCD00063228
Formula:
C₁₃H₁₅NO₄ MW: 249,3 g/mol Melting Pt: 76…78 °C |
MDL Number:
MFCD00063228 CAS Number: 6404-31-5 EINECS: 229-021-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...