Mesitylenesulphonic acid dihydrate ≥98%
Supplier: Thermo Scientific
|
Danger
|
Synonyms:
2,4,6-Trimethylbenzenesulphonic acid dihydrate
B25321.14
B25321.22
ALFAB25321.14EA
0
SGD
ALFAB25321.14
ALFAB25321.22
Mesitylenesulphonic acid dihydrate ≥98%
Mesitylenesulphonic acid dihydrate
Purity: 98+%
CAS No.: 835617-36-2
Molecular Formula: C9H12O3S.2H2O
Formula Weight: 236.29 (200.26anhy)
Melting Point: 74-77°
Beilstein Reference: 1961501
UN No.: UN2585
MDL No.: MFCD00149535
Formula:
C₉H₁₂O₃S·2H₂O MW: 236,29 g/mol (200,26 anhy) Melting Pt: 74…77 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149535 CAS Number: 835617-36-2 EINECS: 222-379-3 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...